missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Trifluoromethylphenylboronic acid, 97+%, Thermo Scientific™

Chemical Identifiers

CAS 1423-26-3
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.93
InChI Key WOAORAPRPVIATR-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylboronic acid, 3-trifluoromethylphenylboronic acid, 3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl benzeneboronic acid, m-trifluoromethyl phenylboronic acid, 3-trifluoromethylboronic acid, 3-trifluoromethyl phenyl boranediol, 3-boronobenzotrifluoride, 3-trifluoromethyl benzeneboronic acid/anhydride
PubChem CID 2734388
IUPAC Name [3-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)C(F)(F)F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359750010
SDS View Documents
Thermo Scientific Chemicals
359750010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC359750050
SDS View Documents
Thermo Scientific Chemicals
359750050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1423-26-3
189.93
3-trifluoromethyl phenylboronic acid, 3-trifluoromethylphenylboronic acid, 3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl benzeneboronic acid, m-trifluoromethyl phenylboronic acid, 3-trifluoromethylboronic acid, 3-trifluoromethyl phenyl boranediol, 3-boronobenzotrifluoride, 3-trifluoromethyl benzeneboronic acid/anhydride
[3-(trifluoromethyl)phenyl]boronic acid
C7H6BF3O2
WOAORAPRPVIATR-UHFFFAOYSA-N
2734388
B(C1=CC(=CC=C1)C(F)(F)F)(O)O

Specifications

1423-26-3
100.0
97% min. (HPLC)
C7H6BF3O2
3-trifluoromethyl phenylboronic acid, 3-trifluoromethylphenylboronic acid, 3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl benzeneboronic acid, m-trifluoromethyl phenylboronic acid, 3-trifluoromethylboronic acid, 3-trifluoromethyl phenyl boranediol, 3-boronobenzotrifluoride, 3-trifluoromethyl benzeneboronic acid/anhydride
B(C1=CC(=CC=C1)C(F)(F)F)(O)O
189.93
189.93
3-Trifluoromethylphenylboronic acid
97.0
Authentic
Glass bottle
1g
WOAORAPRPVIATR-UHFFFAOYSA-N
[3-(trifluoromethyl)phenyl]boronic acid
2734388
97+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only