missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trimethoxysilyl)propyl Acrylate (stabilized with BHT) 93.0+%, TCI America™

Supplier:  TCI America A159725G

Catalog No. A159725G


Explore available promotions

Description

Description

Specifications

Specifications

3-(Trimethoxysilyl)propyl Acrylate (stabilized with BHT)
Yellow
C9H18O5Si
25 g
3-trimethoxysilyl propyl acrylate, 3-acryloyloxy propyltrimethoxysilane, 3-acryloxypropyltrimethoxysilane, 2-propenoic acid, 3-trimethoxysilyl propyl ester, 3-acryloxypropyl trimethoxysilane, 3-trimethoxysilyl propyl 2-propenoate, 3-acryloyloxypropyltrimethoxysilane, 3-acryloxypropyltrimethoxysilane, stab. with 100ppm bht, acmc-1aoz6, acrylic acid, 3-trimethoxysilyl propyl ester
CO[Si](CCCOC(=O)C=C)(OC)OC
234.32
234.32
Liquid
4369-14-6
95°C
MFCD00054803
1760
KBQVDAIIQCXKPI-UHFFFAOYSA-N
3-(trimethoxysilyl)propyl prop-2-enoate
151204
≥93.0% (GC)

Chemical Identifiers

4369-14-6
234.32
KBQVDAIIQCXKPI-UHFFFAOYSA-N
151204
CO[Si](CCCOC(=O)C=C)(OC)OC
C9H18O5Si
MFCD00054803
3-trimethoxysilyl propyl acrylate, 3-acryloyloxy propyltrimethoxysilane, 3-acryloxypropyltrimethoxysilane, 2-propenoic acid, 3-trimethoxysilyl propyl ester, 3-acryloxypropyl trimethoxysilane, 3-trimethoxysilyl propyl 2-propenoate, 3-acryloyloxypropyltrimethoxysilane, 3-acryloxypropyltrimethoxysilane, stab. with 100ppm bht, acmc-1aoz6, acrylic acid, 3-trimethoxysilyl propyl ester
3-(trimethoxysilyl)propyl prop-2-enoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3727

RTECSNumber : UD3810000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions