missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Tropanylindole-3-carboxylate hydrochloride, Thermo Scientific Chemicals
A selective 5-HT3 serotonin receptor antagonist
$413.27 - $413.27
Chemical Identifiers
CAS | 105826-92-4 |
---|---|
Molecular Formula | C17H21ClN2O2 |
Molecular Weight (g/mol) | 320.817 |
MDL Number | MFCD00210221 |
InChI Key | XIEGSJAEZIGKSA-ICHLHHFLSA-N |
Synonym | sdz-ics 930 |
PubChem CID | 71300439 |
IUPAC Name | [(5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 1H-indole-3-carboxylate;hydrochloride |
SMILES | CN1C2CCC1CC(C2)OC(=O)C3=CNC4=CC=CC=C43.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ62807MC
|
Thermo Scientific Chemicals
J62807MC |
100 mg |
Each for $413.27
|
|
|||||
Description
A selective 5-HT3 serotonin receptor antagonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105826-92-4 | |
320.817 | |
XIEGSJAEZIGKSA-ICHLHHFLSA-N | |
71300439 | |
CN1C2CCC1CC(C2)OC(=O)C3=CNC4=CC=CC=C43.Cl |
C17H21ClN2O2 | |
MFCD00210221 | |
sdz-ics 930 | |
[(5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 1H-indole-3-carboxylate;hydrochloride |
Specifications
105826-92-4 | |
C17H21ClN2O2 | |
100 mg | |
14,9783 | |
Soluble in water | |
CN1C2CCC1CC(C2)OC(=O)C3=CNC4=CC=CC=C43.Cl | |
320.817 | |
320.82 | |
3-Tropanylindole-3-carboxylate hydrochloride |
White | |
MFCD00210221 | |
UN2811 | |
sdz-ics 930 | |
XIEGSJAEZIGKSA-ICHLHHFLSA-N | |
[(5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 1H-indole-3-carboxylate;hydrochloride | |
71300439 | |
Powder |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber : NL6012520
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only