missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eagle™ 33 Gallon Red Biohazard Waste Bag
Biohazard waste bag for holding or consolidating larger amounts of biological waste or contaminated materials
Supplier: Eagle™ 1691
Description
- Designed to fit Eagle™ 1601 series drums
- Once bag is full and ready for disposal, seal bag in place and close drum using lid and lever-lock
- Red color bag easily identifiable as biohazard bags
- Drums can be black, yellow, blue or red.
- Printing includes English, Spanish and French translation and international recognizable biohazard symbol
- Material: Linear Low Density Polyethylene (LLDPE)
- Thickness: 1.2 mil
- High level of flexibility and shear resistance can be attained without leaks
- Meets ASTM D 1709-01 and ASTM D 1922-00a specifications
- Tubular construction, single side seal and star seal bottom provide stable, leak proof bag
- 100/Pk.
Specifications
33 gal. | |
100/Pk. | |
LDPE | |
32.5 in. | |
1.2 mil | |
Polyethylene |
Red | |
Not Autoclavable | |
Eagle™ Poly Drums | |
45 in. | |
32.5 in. | |
Biohazard Waste Bags |