Greiner Bio-One 384-well μClear™ Bottom Polystyrene Microplates
Greiner Bio-One 384-Well μClear™ Bottom Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
Supplier:  Greiner Bio-One 781097
 View more versions of this product
Catalog No. 07000053
Explore available promotions
Description
Description
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • Well-to-well spacing: 4.5mm
  • μClear™ bottom
Certifications
DIN ISO 9001
Specifications
Specifications
High Binding
Without Lid
High Binding
Polystyrene
Sterile
Standard
16 x 24
Clear
384
Flat Bottom (F-well)