Greiner Bio-One 384-well μClear™ Bottom Polystyrene Microplates
Greiner Bio-One 384-Well μClear™ Bottom Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
Supplier:  Greiner Bio-One 781096
 View more versions of this product
Catalog No. 07000057
Explore available promotions
Description
Description
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • Well-to-well spacing: 4.5mm
  • μClear™ bottom
Certifications
DIN ISO 9001
Specifications
Specifications
Medium Binding
Without Lid
Medium Binding
Polystyrene
Non-sterile
Flat Bottom (F-well)
Standard
16 x 24
Black
384
Non-treated
40/Cs.