missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ 384-Well Low Flange Polystyrene Microplates
For use with fluorescent assays with detection microplate reader. Corning™ 384-Well Low Flange Polystyrene Microplates have low background fluorescence, minimal light scatter and reduced crosstalk.
Supplier: Corning™ 3769
Description
Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.
Specifications
Standard | |
0.06 cm2 | |
White, Clear Bottom | |
110.5 μL | |
Non-sterile | |
Flat Bottom (F-well) | |
20/Pk. | |
11.6 mm |
With Lid | |
16 x 24 | |
Polystyrene | |
384 | |
Non-binding | |
100/Cs. | |
127.8 x 85.5 x 14.35 mm (5 x 3.36 x 0.56 in.) | |
Top/bottom: 3.63/2.82 mm |