3B Scientific™ Biochemistry Molecular Model Kit
Compact/semi-space-filling models feature the mushroom style hydrogen atom-link part.
Supplier:  3B Scientific™ ABW19720
Catalog No. S68292
Explore available promotions
Includes:
  • 68 carbon, black (42 tetrahedral, 24 trigonal, two linear)
  • 34 nitrogen, blue (12 tetrahedral, 12 trigonal, 10 angular)
  • 40 oxygen, red (20 angular, 10 linear, 10 one-hole)
  • 10 hydrogen, white (linear)
  • Two sulfur, yellow (angular)
  • Six phosphorus, purple (tetrahedral)
  • Two metal connectors, gray (one tetrahedral, one octahedral)
  • 100 hydrogen links, white
  • 150 NV-links, clear
  • 10 V-links, gray
  • Two link remover tools, cream
Description
Description
257 atom parts cover a wide range of biochemical structures including amino acids, monosaccharides, fatty acids, glycerol, steroids, purines, pyrimidines, peptides, disaccharides, nucleosides, nucleotides, coenzymes, proteins, polysaccharides and nucleic acids.
Specifications
Specifications
Molecular Model, Biochemistry Kit
68 Carbon, black (42 tetrahedral, 24 trigonal, two linear), 34 Nitrogen, blue (12 tetrahedral, 12 trigonal, 10 angular), 40 Oxygen, red (20 angular, 10 linear, 10 one-hole), 10 Hydrogen, white (linear), Two Sulfur, yellow (angular), Six Phosphorus, purple (tetrahedral), Two Metal connectors, gray (one tetrahedral, one octahedral), 100 Hydrogen links, white, 150 NV-links, clear, 10 V-links, gray, two link remover tools, cream