missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Thermo Scientific™ 3D Tank Liner, ASI™ 26 film
Single-use tank liners reduce cleaning steps common to re-usable tanks.
$807.18 - $2704.38
Specifications
Material | Polyethylene |
---|---|
Content And Storage | Store at room temperature. |
Outer Packaging | Individually packed in a header bag |
Sterilization Method | Irradiated |
Shipping Condition | Room Temperature |
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Dimensions | Price | Quantity | ||||
B100363I
|
Thermo Scientific™
B100363I |
200 L | 55.9 cm x 91.4 cm (22 in. x 36 in.) |
Case of 20 Each for $1,469.40
|
|
||||
B100364I
|
Thermo Scientific™
B100364I |
50 L | 33 cm x 68.6 cm (13 in. x 27 in.) |
Case of 40 Each for $2,704.38
|
|
||||
B100373I
|
Thermo Scientific™
B100373I |
100 L | 45.7 cm x 76.2 cm (18 in. x 30 in.) |
Case of 20 Each for $1,206.14
|
|
||||
B101459I
|
Thermo Scientific™
B101459I |
300 L | 70 cm x 122 cm (24 in. x 48 in.) |
Case of 10 Each for $807.18
|
|
||||
B101460I
|
Thermo Scientific™
B101460I |
560 L | 78.7 cm x 124.5 cm (31 in. x 49 in.) |
Case of 10 Each for $871.69
|
|
||||
Description
The Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
- Minimizes bioburden due to construction under sterile cGMP conditions
- Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Polyethylene | |
Individually packed in a header bag | |
Room Temperature | |
0 Ports | |
No Tubing |
Store at room temperature. | |
Irradiated | |
HyPerforma™ | |
ASI 26 |