3M™ Disposable Protective Coveralls 4520, Hooded
Coveralls protect against certain light liquid splashes and hazardous dusts while enhancing movement and comfort.
$193.12 - $206.65
Specifications
Product Type General Purpose Coveralls
Gender Unisex
Material Microporous Polyethylene Film
Color White
Disposable Single-use
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-623-346
View Documents
3M™
7000089644
4X-Large
Case of 25 Each for $206.65
17-623-345
View Documents
3M™
7000089643
3X-Large
Case of 25 Each for $193.12
Description
Description
  • Designed for use in industrial workplaces as a physical barrier to help prevent skin contact from liquid splashes and dust in certain work environments
  • Seamless shoulder and sleeve tops help provide increased comfort and fewer potential entry points for contaminants
  • Meet ANSI 101-1996 size guidelines
  • Meets EN 340, Type 6 Splash - EN 13034/(EN ISO 17491-4), Type 5 Dust - EN ISO 13982-1, Anti-Static, EN 11492 and Nuclear - EN 1073 - 2
  • Microporous PE laminate
  • Breathable, SMS fabric back panel helps improve air ventilation and reduce heat build-up
  • Reinforcement gusset between the legs to help provide increased durability during bending and squatting
  • Elastic waist and ankles, knit cuffs, two-way zipper with sealable storm flap
  • Hooded style for use with respiratory equipment when extra protection is required
Specifications
Specifications
General Purpose Coveralls
Microporous Polyethylene Film
Single-use
Elastic
Hooded
Front Zipper with Storm Flap
Non-sterile
Anti-static
CE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073
No
Insulation Installation, Asbestos and Lead Abatement, Coal Dust in Power Plants, Woodworking, Metal Polishing, Light-Duty Building Cleaning, General Powder Handling, General Industrial Clean-up and Maintenance, Machine or Vehicle Maintenance and Food Processing
Unisex
White
Elastic
Elastic, Knit
3-panel
Serged
Dust, Liquid
ANSI 101-1996
3 Layers of Melt-blown Polypropylene in-between 2 layers of Spunbound Polypropylene
4520
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'