missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ 4530 Series Disposable Protective Coveralls
The 3M™ Protective Coverall 4510 is disposable safety work wear made of a high quality microporous laminated material. It helps provide basic barrier protection against light liquid splashes and hazardous dusts.
Supplier: 3M™ 4530EXL
Description
The key features make this protective coverall a great disposable PPE option. The 3M protective coverall 4530 helps protect against dusts and light liquid splashes. Typical applications for this safety work wear may include: Welding, metal grinding, General Powder Handling, and Woodworking. Designed to be worn over primary thermal protective FR garments. Product recommended for the following industries: Abatement/Remediation, Agriculture, Cleaning and Maintenance, General Industry/Manufacturing, Non-Hazardous Operations, Welding/Metalwork.Specifications
General Purpose Coveralls | |
Unisex | |
Blue | |
Elastic, Knit | |
Non-sterile | |
CE, EN 1149-5:2008, EN 1073-2:2002 | |
43 to 45 in. | |
Cement Manufacturing, Polishing, Oils and Petrochemicals, Glass Fibre, Light Industrial Cleaning, Painting and Coating, Resins | |
179 to 187 cm |
X-Large | |
Polypropylene | |
Single-use | |
Front Zipper with Storm Flap | |
Anti-static | |
Blue SMMMS Polypropylene suit and white back panel, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester cuffs and thread | |
109.22 to 114.3 cm | |
5.83 to 6.16 ft. |