3M™ Powerflow™ High Efficiency SP3 Particulate Filter
High Efficiency (HE) SP3 particulate filter system component is designed for use with Powerflow™ PAPR systems.
Supplier:  3M™ 7000131701
Catalog No. 29-635-7476
Explore available promotions
Description
Description
  • 3M Catalog# 024-00-02R01 , 3M Catalog# 450-01-01R20
Note: These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Specifications
NIOSH
Sp3 HEPA Filter
Powerflow™ PAPR Systems
Magenta
Ideal to protect against dusts, mists, fumes, asbestos, radionuclides and radon daughters
450-01-01R20