3M™ Air-Mate™ High-Efficiency Belt-Mounted PAPR System Breathing Tube
Replacement part for Air-Mate™ PAPR system.
Supplier: 3M™ 7000005588
Description
- Motorized systems powered by battery pack that draws air through filter or cartridge
- Lower unit can be mounted on waist, face, or head, depending on system selected
- Back-Mounted
- Constant air flow provide cooling for worker that results in greater comfort for wearer
- Loose-fitting headgear options provide respiratory protection without need for tight faceseal or fit testing
- Battery-powered blower pulls air through filter resulting without additional breathing resistance to wearer
- 3M Catalog# BE-224
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Black | |
Breathing Tube Assembly | |
Air-Mate™ PAPR systems | |
BE-224 |