3M™ Belt-Mounted and Vehicle-Mounted PAPR System Accessory, Cartridges
Choose from a large selection of cartridges for powered air purifying respirators used in a variety of industrial environments.
Supplier:  3M™ 7000029620
 View more versions of this product
Catalog No. 19-087394
Explore available promotions
Description
Description
  • Cartridge attaches to the motor blower
  • Filters supply air to PAPR headgear of choice
  • Products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product
  • 3M Catalog# GVP-440, GVP-441
Specifications
Specifications
Organic Vapor, Acid Gas, High Efficiency (HE)
Magenta/Yellow
Transportation, Mining, Oil & Gas, General Manufacturing
GVP-443
NIOSH
Cartridge
Belt-mounted and vehicle-mounted PAPR systems
6/Cs.