3M™ Cartridges for Full-Facepiece FR-M40
Reduce exposure to chemical and biological warfare agents
Supplier: 3M™ 7000052356
Description
Cartridge-type filter media compatible with FR-M40 facepieces. (See also FR-64 cartridge [Cat. No. 19-813-754], listed separately.)
FR-C2A1 Cartridge
- Contains a carbon bed filter, tested to MIL-C-51560A (EA) and EA-C-1704 specifications with hydrogen cyanide, cyanogen chloride, phosgene, dimethyl methylphosphonate (a nerve agent stimulant) and sarin (GB)
- Contains a particulate filter, tested to be at least 99.97% efficient against a 0.3mm MMAD (Mass Median Aerodynamic Diameter) aerosol challenge at 85 L/min.
- Designed to help provide respiratory protection against certain airborne contaminants when used in accordance with all use and limitation instructions and applicable safety and health regulations
- 3M Catalog# FR-15-CBRN
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Certifications
NIOSH approved with FR-M40 facepiece for sulfur dioxide, chlorine, hydrogen chloride, CN, CS and P100 particulate.
Specifications
CBRN | |
Canister | |
Fr-m40, 6000, and 7800s series negative pressure facepieces | |
4/Cs. |
NIOSH | |
Commonly found in law enforcement, fire and emergency response (paramedics) environments | |
FR-15-CBRN |