missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Disposable Protective Coverall 4530
Disposable coveralls offer protection against certain light liquid splashes and hazardous dust. They feature a low-profile, three-panel hood that zips up to the chin for additional head protection.
Supplier: 3M™ 7000089024
Description
- Breathable material (SMMS) to help reduce heat build up
- Elastic waist and ankles for convenience and freedom of movement
- Knit cuffs for increased comfort
- Standard collar provides convenience when using auxiliary PPE
- Two-way zipper with sealable storm flap for convenience
- Flame retardant coating offering limited protection against heat and flame
- Reinforced gusset
- Anti-static treatment on both sides
- Protects against dusts and light liquid splashes
- Typical applications may include welding, metal grinding, general powder handling, and woodworking
- Designed to be worn over primary thermal protective FR garments
Specifications
General Purpose Coveralls | |
Unisex | |
Blue | |
Elastic | |
Knit | |
Front Zipper | |
Non-sterile | |
Standard Fold | |
Suit: SMS Polypropylene, Back panel: SMS Polypropylene, Zipper: Nylon on Polyester Braid, Elastic: Neoprene Rubber, Cuffs: Polyester, Thread: Polyester | |
4530CS | |
32 in. | |
100 to 108 cm | |
Construction, Mining, Oil and Gas, Abatement, Remediation, Cleaning, and Maintenance | |
174 to 181 cm |
Large | |
SMS Fabric | |
Single-use | |
Elastic | |
Collared | |
Serged, Bound | |
Anti-static | |
EN 13034, EN 13982, EN 1149, EN 1073, ISO 14116 | |
No | |
39 to 43 in. | |
51 in. | |
129 cm | |
69 to 71 in. |