Learn More
3M™ E-A-R™ Express™ Corded Metal Detectable Ear Plugs
E-A-R™ metal detectable ear plugs have a stainless steel bead encased in the ear plug. Popular in food manufacturing industries, when contamination prevention is critical.
Supplier:  3M™ 7000127208
Catalog No. 29-635-1076
Only null left
Explore available promotions
Description
Description
  • Implanted stainless steel ball bearings aid in locating lost ear plugs
  • Designed for food processing and other industries
  • Paddle-style grips provide quick, clean, and consistent placement of ear plugs
  • No-roll design keeps fingers, ear plugs, and ears clean
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Yes
Class AL
Food Processing, Manufacturing, Pulp and Paper, Pharmaceutical, Tobacco Processing Industries and Other Various Industries That Require Detectable Products to Prevent Contamination
311-1127
400 Pairs/Cs
Yellow
Corded
Reusable
Blue
Universal
Yes
Poly bag
Foam, thermoplastic
26 dB
Pod
Yes
Product Suggestions
Product Suggestions