3M™ E-A-Rsoft™ FX™ Ear Plugs
FX™ ear plugs feature a bell shape and super-soft, slow-recovery foam to effectively seal the ear canal and help provide reliable hearing protection. Flared end helps make fitting and removal easier.
Supplier:  3M™ 7000029951
 View more versions of this product
Catalog No. 19-153-650
Explore available promotions
Description
Description
  • Soft, pliable foam offers comfort for wearing over an extended time
  • Bell shape allows easy fitting and removal
  • Slow recovery foam effectively seals the ear canal and provides reliable hearing protection
  • Test compatible with 3M™ E-A-Rfit™ Dual-ear Validation System
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
Yes
Universal
Yes
Poly bag
Foam
33 dB
Bell
No
Class AL
Assembly, Blasting, Cleaning, Composite Finishing, Demolition, Electrical, Facility Maintenance, Final Finish, Grinding, Machine Operations, Paint Preparation, Painting, Sanding, Welding
312-1261
2000 Pairs/Cs
Yellow
Uncorded
Reusable