3M™ Ethylene Oxide Monitor
No batteries, hoses, pumps or equipment to calibrate
Supplier:  3M™ 7000051749
 View more versions of this product
Catalog No. 17-985-363B
Explore available promotions
Description
Description
  • Ideal for chemical, medical, and pharmaceutical labs.
  • Sensitivity of 0.24ppm-hours
  • Upper exposure limit of 600ppm-hours
  • Available as a monitor only or with prepaid analysis
  • These safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
  • 3M Catalog# 3551
Misuse may result in sickness or death.
Specifications
Specifications
Ideal for Chemical, Medical, and Pharmaceutical Labs
Ethylene Oxide
Ethylene Oxide Monitor only
OSHA
600 ppm.Hour
3551