3M™ FR-7800B Respirator Full Facepieces
Integral speaking diaphragm improves voice amplification and clarity
Supplier:  3M™ 7000126539
 View more versions of this product
Catalog No. 19-130-4853A
Explore available promotions
Description
Description
  • Features a unique double flange face sealing surface that enhances fit and protection
  • Designed for first responder applications commonly found in law enforcement, fire, emergency response and medical environments
  • Canister can be mounted on either side to accommodate different weapon sighting styles
  • Lens has optical correction promoting no left-to-right visual distortion
  • 3M Catalog# FR-7800B-L , 3M Catalog# FR-7800B-M , 3M Catalog# FR-7800B-S
Certifications
NIOSH approved to Chemical Biological Radiological Nuclear (CBRN) requirements when used with the 3M Canister FR-15-CBRN (19-130-0673).
Compliance
Meet ANSI™ Z87.1-2003 standards.
Specifications
Specifications
NIOSH, CBRN, ANSI Z87.1-2003
Full Facepiece
CBRN applications when used with 3M™ canister fr-15-CBRN
Integral speaking diaphragm
6-point strap
Silicone, Butyl Rubber Faceshield
Small
Air purifying respirator
FR-7800B-S