3M™ Full Facepiece 6000DIN Series Respirator
Lightweight full-facepiece respirator with built-in DIN port adapter assembly features a well-balanced design and silicone faceseal for enhanced comfort, durability and ease of cleaning. Available in air-purifying, supplied air, and powered air purifying respirator modes.
Supplier:  3M™ 7000029682
 View more versions of this product
Catalog No. 19-055-520
Explore available promotions
Alert:
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Requires : Filter media (sold separately).
Description
Description
  • Can be reconfigured for use with 2000 Series or 7093 filters, 6000 Series cartridges, and powered or supplied air systems against a variety of gases, vapors, and particulate hazards according to NIOSH approvals
  • Soft, lightweight, and well-balanced design
  • Adjustable head straps for long-wearing comfort
  • Large polycarbonate lens with scratch-resistant coating
  • No components made from natural rubber latex
  • 3M™ Cool Flow™ Valve helps make breathing easier
  • Available in air-purifying, supplied air, and powered air purifying respirator modes
  • Center adapter directs exhaled breath downward
Order Info
Facepieces and responder filter media sold separately. Consult your Fisher Safety Sales Representative for more information about appropriate uses and configurations.
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Compliance
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters.
Specifications
Specifications
ANSI Z87.1
Bayonet
Silicone, Thermoplastic Elastomer
Medium
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
Din port adapter assembly
6800DIN
NIOSH 42 CFR Part 84
Gray
Gas Mask Respirator
Silicone
3M™ Filters 2000 Series, 3M™ Filter 7093, P100, 3M™ Cartridges 6000 Series
Polycarbonate
4-point