3M™ H10 Ratchet Headgear
Ratchet headgear with crown extender for use with any 3M™ Faceshield helps provide face protection. High-strength thermoplastic crown provides impact resistance. Adjustable ratchet suspension with five position crown strap.
Supplier: 3M™ 7000052751
Description
Features a unique system that allows faceshield window replacement in seconds and helps provide a precise fit and comfortable head protection.3M provides reliable head and face protection products that meet regulated performance and safety requirements.- Adjustable ratchet suspension
- Crown extender for added coverage
- Five position crown strap
- Headgear gives users a wide choice of faceshield materials, sizes, shapes, and tints for task specific requirements
- Meets the requirements of ANSI Z871-2003
- 3M Catalog# 82516-00000