missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ Peltor™ Bluetooth™ Headsets

Two-way communication headset for industrial users in high-noise environments

Supplier:  3M™ MT53H7AWS2

Catalog No. 19-130-3645


Explore available promotions

Description

Description

  • Freedom to roam up to 32 ft. from mobile radio with wireless Bluetooth™
  • Comfortable for extended use, with soft cushions and deep earcup design
  • Convenient power with two “AA” batteries, or rechargeable Ni-MH battery pack
  • Protect hearing with Noise Reduction Rating of 25 dB

Headband Headset

  • Over-the-head Bluetooth communications headset provides clear, concise wireless communications, plus reliable hearing protection compatible with most two-way radio systems
Neckband and Hardhat headsets
  • Convenient, easy-to-use neckband Bluetooth communications headset is part of 3M Peltor Wireless Solutions™ series, which offers wireless connection, ability to synch to an existing communication system and hearing protection

Heavy equipment operators; Airport workers; Construction; Manufacturing; Agricultural; Racing

Specifications

Specifications

Soft cushions and deep earcup design
Headband style
Bluetooth Headset
Roam up to 32 ft. from mobile radio with wireless Bluetooth
25 dB
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions