3M™ 3M™ PELTOR™ Boom Microphone
Enables clear communication when using a 3M™ Versaflo™ M-300 Series Respirator Hard Hat. Compatible with the MT53 series 3M™ PELTOR™ Protective Communication Headsets. This noise-cancelling speech microphone enables clear communication in noisy environments.
$217.10 - $217.10
Specifications
Product Type Microphone
Quantity 20 /Cs.
View More Specs
Products 1
Catalog Number Mfr. No. Model Price Quantity  
Catalog Number Mfr. No. Model Price Quantity  
03-051-108
View Documents
3M™
7100099097
MT7V/1
Case of 120 Each for $217.10
Description
Description
  • Noise-cancelling microphone for clear communication in noisy environments
  • For use with the 3M™ Versaflo™ M-300 Series Hard Hat
  • Compatible with 3M™ PELTOR™ Headsets that have a product number starting with MT53 or MRX21
  • Microphone includes three adjustment points to fit to a wide range of head sizes
  • Easily snaps into the rim of the hard hat
  • Hard hat attachment plates are included for the assembly of a 3M™ PELTOR™ Headset to the hard hat
  • This accessory is included in the NIOSH approvals for M-300 series configurations
Specifications
Specifications
Microphone
20 /Cs.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'