3M™ Privo™ Safety Eyewear
Offers European styling
Supplier: 3M™ 7000052818
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
- Lens and temple tip color options
- Soft-tipped temples
- Soft comfort nose bridge
- Durable polycarbonate lenses absorb 99.9% UV
- 3M Catalog# 12261-00000-20, 12263-00000-20, 12264-00000-20, 12265-00000-20, 12266-00000-20, 12268-00000-20
Compliance
Meets ANSI™ Z87.1-2010 and CSA Z94.3 standards.
Specifications
Meets ANSI Z87.1-2010 and CSA Z94.3 standards. | |
Black | |
Polycarbonate |
Amber | |
12263-00000-20 |