3M™ Ratchet Headgear, Head and Face Protection with Chin Protector
Headgear and chin protector assembly helps deliver reliable face protection. Visor not included
Supplier: 3M™ 7000002293
Alert:
Protective eyewear should be worn under faceshields at all times and is sold separately.
Description
- High-strength thermoplastic crown protector provides impact resistance
- Can be used when extra chin protection is needed
- Clear chin protector allows downward visibility
- With adjustable ratchet suspension, this headgear features a unique system that allows faceshield window replacement in seconds and helps provide a precise fit and comfortable head protection
- 3M Catalog# 82521-10000
Order Info
Uses 3M™ Faceshield WP98 (82543-0000); available separately, see Cat. No. 19-070-902.
Specifications
ANSI Z87.1-200 | |
Head and Face Combinations Protection | |
High-strength ratchet headgear and clear polycarbonate chin protector (Fisher Cat: 19-040-042) | |
Thermoplastic |
Ratchet | |
Blue | |
82521-10000 |