3M™ Rugged Comfort 6581/55886 Head Harness Assembly
The 3M™ Rugged Comfort Respirator is designed with harsh, dirty environments in mind. Offers comfort, durability and stability with a firm, lightly textured silicone faceseal and a low profile, strong unibody construction.
Supplier:  3M™ 7000128251
Catalog No. 19-096-917
Explore available promotions
Description
Description
Rugged and Comfortable Respiratory Protection, the 3M™ 6500 Reusable Respirator
  • Individual packaging designed for vending
  • Protects products in vending environment and process
  • NIOSH P95 filter media helps protect against certain oil and non-oil based particles
  • Carbon layer reduces exposure to nuisance levels* of organic vapors
  • 3M™ CoolFlow™ Valve and adjustable M nose clip
  • Braided headbands and soft inner materials
  • Features Include:Silicone face seal
    • Provides comfort and stability with a soft, but firm face seal
    • Provides long life with a resilient silicone material
    • Our lightest silicone facepiece
    • Keep its shape under high heat environments
    • Quick Latch - Drop Down Mechanism
      • Easy on and off as you move in and out of contaminated areas
      • No need to remove your hard hat or face shield when lowering or raising your respirator
      • Adjustable Head Harness Assembly
        • Optimum fit and comfort with four point adjustable cradle
        • Lasting performance and improved elasticity Polyester/Spandex straps
        • Overmolded/Low Profile Design
          • Simplified cleaning and maintenance with fewer parts and crevices
          • A good fit on many faces with a smaller face seal footprint
          • .
          • Improved compatibility for use with Welding/Grinding Shields
          • Advanced half facepiece design for a wide field of view
          • 3M™ Bayonet Connection
            • Compatible with all 3M™ cartridges and filters
            • Coolflow™ Valve
              • Enhanced comfort by reducing heat and moisture
              • Easier breathing with unique valve design
              • Valve Cover design
                • Directs exhaled breath and moisture downward
                • Easy positive pressure seal check
                • 3M Catalog# 6581
Specifications
Specifications
3M 6500 Series