3M™ Versaflo™ S-Series Hood with Faceseal, Integrated Head Suspension
Versaflo™ S-Series Hood with integrated suspension provides head, face, neck, and shoulder coverage as well as providing eye and face protection from liquid splash and low energy flying particles and dust.
Supplier:  3M™ 7000127431
Catalog No. 19-805-615
Explore available promotions
Description
Description
  • Integrated head suspension provides head, face, neck and shoulder coverage.
  • Protects face and eyes from liquid splash and low energy flying particles and dust (Grade F).
  • Suitable for applications which require frequent replacement of the entire headtop assembly or those where there is less soiling of the outer fabric.
  • No assembly required
  • Ready to use straight out of the box.
Specifications
Specifications
1000
White
Hood with Faceseal
Assembly, facility maintenance, machine operation, painting, sanding
PETG
5/Cs.
19.88 in.
10.63 in.
9.06 in.
NIOSH
Polypropylene
Small/Medium
GVP, Breathe-Easy™, V Series, T-300, T-600, T-800
S-433S-5
19.88 x 10.63 x 9.06 in. (50.49 x 27 x 23.01 cm)
50.49 cm
27 cm
23.01 cm