missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Scotch™ Box Sealing Tape 371
Dependable sealing performance
$6.72 - $6.72
Specifications
Description | Premium |
---|---|
For Use With (Application) | Carton sealing tape, box sealing tape and box closing tape |
Model | 371 |
Catalog Number | Mfr. No. | Color | Dimensions (LxW) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Dimensions (LxW) | Price | Quantity | |||||
19-520-018
|
3M™
7100011547 |
Clear | 100 m x 48 mm |
Each for $6.72
Case of 36 Each for $241.92
|
|
|||||
Description
- Anindustrial packaging tape used for box sealing, splicing, recouperage and other packaging applications
- Performs well on single-wall, low burst-strength regular slotted containers
- Biaxally oriented 1.9mil polypropylene with film backing resists score line breaking and center seam splitting
- Pressure-sensitive adhesive adheres instantly to a variety of surfaces
- 3M Catalog# 371, 371, 371, 371
Specifications
Premium | |
Carton sealing tape, box sealing tape and box closing tape | |
371 |