3M™ Versaflo™ Easy Clean TR-300N+ ECK PAPR Kit
3M™ Versaflo™ TR-300N+ ECK is a complete, ready to use HE PAPR system that provides protection against particulates. Easy to clean and maintain.
Supplier:  3M™ 7100153814
Catalog No. 17-000-724
Explore available promotions
Description
Description
Features:
  • Conveniently disposable - Save time cleaning with our maintenance-free disposable hood
  • Wider field of vision - Freedom to wear your own safety eyewear
  • Connect quickly - Our innovative Quick Release Swivel (QRS) connects quickly and helps avoid kinks in the breathing tube
  • Contoured design - Slim device with smooth outer surfaces can help work in tight spaces
  • Two selectable flow rate options offers more options and comfort to the user
  • Easy-to-use and versatile PAPR system for particulate environments
  • High Efficiency particulate filter. Optional nuisance organic vapor filter available
  • Suggested industries include: Pharmaceutical, Healthcare and Food Safety
  • 3M Catalog# TR-300N+ECK
Certifications
NIOSH Approved
Compliance
NIOSH approved.
Specifications
Specifications
10 to 12 hours
NIOSH
HEPA
6.5 CFM and 7.2 CFM
S-433L Hood w/ Integrated Hood Suspension, BT-30 Length Adjusting Breathing Tube, TR-302N+ PAPR Unit, TR-327 Easy Clean Belt, TR-332 High Capacity Battery, TR-341N Battery Charger Kit, TR-3712N HE Filter, TR-3600 Pre-Filter, TR-971 Airflow Indicator
Belt-mounted
High Capacity Lithium Ion
Beige
PAPR System with Hood
Powder Handling
TR-300N+ECK