3M™ Versaflo™ Heavy Industry TR-300N+ HIK PAPR Kit
The 3M™ Versaflo™ TR-300-HIK is a complete, ready to use HE PAPR system for industrial applications. Provides protection against particulates.
Supplier:  3M™ 7100153815
Catalog No. 17-000-725
Explore available promotions
Description
Description
Features:
  • Designed for compatibility - easily accommodate eyeglasses and attach earmuffs
  • Beat harsh environments - Rugged work demands durable and strong head protection
  • See clearer - Durable faceshield helps protect against flying objects and sparks while retaining optical clarity
  • Breathe easier - When working in hot, humid conditions, stay comfortable with powered air
  • Work longer - the High Efficiency battery allows you to work in a remote location for extended periods of time
  • Two selectable flow rate options offers more options and comfort to the user
  • Easy-to-use and versatile PAPR system for particulate environments
  • High Efficiency particulate filter.
  • 3M Catalog# TR-300N+HIK
Certifications
NIOSH Approved
Compliance
NIOSH approved.
Specifications
Specifications
10 to 12 hours
NIOSH
HEPA
6.5 CFM and 7.2 CFM
TR-300N+HIK
High Capacity Lithium Ion
Beige
PAPR System for Heavy Industry
Powder Handling
Belt-mounted