3M™ Versaflo™ M-400 Series PAPR Outer Shroud
Outer shrouds are replacement parts for use with M-400 helmets.
$86.32 - $148.58
Specifications
Assigned Protection Factor (APF) | 1,000 |
---|---|
Product Type | Outer Shroud |
For Use With (Application) | Cleaning, electrical, facility maintenance, painting, sanding, welding |
For Use With (Equipment) | M-400 helmet |
Catalog Number | Mfr. No. | Style | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Style | Price | Quantity | |||||
19-039-526
|
3M™
7100289903 |
Standard |
Case for $86.32
|
|
|||||
19-039-527
|
3M™
7000127685 |
Comfort |
Case for $127.67
|
|
|||||
19-039-529
|
3M™
7100289940 |
High Durability |
Case for $148.58
|
|
|||||
Description
- The M-446 Outer Shroud is a comfortable, low-linting outer shroud for 3M™ Versaflo M-400 Helmets
- Made from Nomex, the M-447 Outer Shroud is a flame resistant outer shroud for 3M™ Versaflo M-400 Helmets
- Made from Cordura Nylon, the M-448 Outer Shroud is a high durability outer shroud for 3M™ Versaflo M-400 Helmets
- 3M Catalog# M-445, M-446, M-447, M-448
Specifications
1,000 | |
Outer Shroud | |
Cleaning, electrical, facility maintenance, painting, sanding, welding | |
M-400 helmet |