3M™ Versaflo Painter's Supplied Air Respirator Kit
The SA-100-PSK consists of a lightweight respiratory face shield, breathing tube, and vortex cooling valve. A Supplied Air Hose (sold separately) is required to complete the NIOSH-approved system. Ideal as a painter's respirator kit.
Supplier:  3M™ 7000127696
Catalog No. 19-039-550
Explore available promotions
Description
Description
The SA-100-PSK includes a V-100 Vortex Cooling Assembly, a BT-20L Breathing Tube with BT-922 Breathing Tube Cover, and a M-101 Respiratory Faceshield with a M-925 Visor and M-936 Comfort Faceseal.
  • Use 3M™ PELTOR™ Push-To-Talk (PTT) Adapters to optimize the placement of the PTT button for convenient activation
  • For use with 3M™ PELTOR™ COMTAC™™ Protective Communication Headsets and compatible two-way radios
  • Ambidextrous design allows for easy use by either the right or left hand
  • Mounting clip can be rotated 360 degrees
  • Design allows for either belt or shoulder mounting
  • Button profile and tactile feedback suitable for gloved hands
  • Optional shroud protects PTT button from accidental activation, for instance when laying in the prone position
  • NEXUS TP-120 connection serves as a break-away connector for removal in emergency situations
  • Curly cable downlead with radio specific connector To determine the appropriate PTT adapter for your two-way radio, please visit 3Mcom/PeltorAdapters
  • NATO wired
  • 3M Catalog# SA-100-PSK
Specifications
Specifications
NIOSH
Painter's Supplied Air Respirator Kit
V-100 vortex cooling assembly, a bt-20l breathing tube with bt-922 breathing tube cover and a m-101 respiratory faceshield with a m-925 visor and m-936 comfort faceseal
SA-100-PSK