3M™ Versaflo™ TR-800-ECK Easy Clean PAPR Assembly Kit
3M™ Versaflo™ TR-800-ECK is a complete Powered Air Purifying Respirator featuring an Intrinsically Safe motor/blower and everything required to to easily clean the PAPR system.
Supplier:  3M™ 7100150926
Catalog No. 17-000-736
Explore available promotions
Includes: Hood, Integrated Head Suspension, Breathing Tube, HE Filter, Battery Charger, Tube Cover, IS PAPR Unit, IS Battery, Battery Attachment Tool, Easy Clean Belt, Filter Cover, Prefilter, Spark Arrestor, Storage and Cleaning Kit, Airflow Indicator
Description
Description
Features:
  • Top of the line IS protection - The TR-800 is a PAPR blower rated at Division I for Classes I,II, and III under the most current UL standard (UL 60079, 6th Edition, 2013)
  • See the bigger picture - Maintenance-free disposable hood saves you time cleaning while providing a wide field of view.
  • More options for more tasks - Choose from particulate filters or gas and vapor combination cartridges for protection in a variety of environments.
  • Work at your own speed - Multiple airflow rates put you in control of your comfort.
  • When clean is critical - Easy-to-clean belt saves time and effort, and storage and cleaning kit pave the way for more productivity.
  • 3M Catalog# TR-800-ECK
Certifications
NIOSH Approved
Compliance
UL-certified Intrinsically Safe for Division 1: Class I, II, III; Division 1 (includes Division 2) Groups C, D, E, F, G; T4, under the most current standard (UL 60079, 6th Edition, 2013).
Specifications
Specifications
6 to 10 hours
UL-Certified Intrinsically Safe for Division 1: Class I, II, III, Division 1 (includes Division 2) Groups C, D, E, F, G, T4 (UL 60079, 6th Edition, 2013)
PAPR System with Hood
TR-800-ECK
Lithium-ion
HEPA
Powder Handling; Liquid Chemical Handling; Powder Coating; Mixing; Painting; Cleaning; General Maintenance; Spray Painting
Belt-mounted
Product Suggestions
Product Suggestions