3M™ Virtua™ Sport Eyewear
Sport styling with maximum protection
Supplier:  3M™ 1154200000
 View more versions of this product
Catalog No. 19-810-008
Explore available promotions
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
  • Polycarbonate lens absorbs 99.9% UV
  • Nine-base curve lens for wrap and front/side coverage
  • Sleek unisex styling in a comfortable, light polycarbonate frame
  • 3M Catalog# 11384-00000-20, 11385-00000-20, 11386-00000-20, 11388-00000-20, 11542-00000-20, 11543-00000-20, 11544-00000-20, 11743-00000-20
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
Anti-fog
Light Blue
11542-00000-20
ANSI Z87.1-2015, CSA Z94.3-2010
Blue
Polycarbonate (High-impact)