missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(1,2,3-Thiadiazol-4-yl)benzylamine hydrochloride, ≥95%, Thermo Scientific™

$185.91 - $185.91

Chemical Identifiers

CAS 175205-49-9
Molecular Formula C9H10ClN3S
Molecular Weight (g/mol) 227.71
MDL Number MFCD02682075
InChI Key WUUGGOQPXZDQNW-UHFFFAOYSA-N
Synonym 4-1,2,3-thiadiazol-4-yl benzylamine hydrochloride, 4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 1-4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 4-thiadiazol-4-yl phenyl methanamine hydrochloride, benzenemethanamine,4-1,2,3-thiadiazol-4-yl, 4-1,2,3-thiadiazol-4-yl phenyl methylamine, chloride, 4-4-aminomethyl phenyl-1,2,3-thiadiazole hydrochloride
PubChem CID 2737287
IUPAC Name [4-(thiadiazol-4-yl)phenyl]methanamine;hydrochloride
SMILES C1=CC(=CC=C1CN)C2=CSN=N2.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC16413CB
SDS View Documents
Thermo Scientific Chemicals
CC16413CB
250 mg Amber Glass Bottle
Each for $185.91
 
Description

Description

Specifications

Chemical Identifiers

175205-49-9
227.71
WUUGGOQPXZDQNW-UHFFFAOYSA-N
2737287
C1=CC(=CC=C1CN)C2=CSN=N2.Cl
C9H10ClN3S
MFCD02682075
4-1,2,3-thiadiazol-4-yl benzylamine hydrochloride, 4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 1-4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 4-thiadiazol-4-yl phenyl methanamine hydrochloride, benzenemethanamine,4-1,2,3-thiadiazol-4-yl, 4-1,2,3-thiadiazol-4-yl phenyl methylamine, chloride, 4-4-aminomethyl phenyl-1,2,3-thiadiazole hydrochloride
[4-(thiadiazol-4-yl)phenyl]methanamine;hydrochloride

Specifications

175205-49-9
100.0
Amber Glass Bottle
MFCD02682075
4-1,2,3-thiadiazol-4-yl benzylamine hydrochloride, 4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 1-4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 4-thiadiazol-4-yl phenyl methanamine hydrochloride, benzenemethanamine,4-1,2,3-thiadiazol-4-yl, 4-1,2,3-thiadiazol-4-yl phenyl methylamine, chloride, 4-4-aminomethyl phenyl-1,2,3-thiadiazole hydrochloride
C1=CC(=CC=C1CN)C2=CSN=N2.Cl
227.71
227.72
4-(1, 2, 3-Thiadiazol-4-yl)benzylamine hydrochloride
94.5
95%
C9H10ClN3S
250 mg
WUUGGOQPXZDQNW-UHFFFAOYSA-N
[4-(thiadiazol-4-yl)phenyl]methanamine;hydrochloride
2737287
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'