missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(1-Propenyl)-1,2-dimethoxybenzene 98.0+%, TCI America™
$43.13 - $263.78
Chemical Identifiers
CAS | 93-16-3 |
---|---|
Molecular Formula | C11H14O2 |
Molecular Weight (g/mol) | 178.231 |
MDL Number | MFCD00009282 |
InChI Key | NNWHUJCUHAELCL-PLNGDYQASA-N |
Synonym | 1,2-Dimethoxy-4-(1-propenyl)benzene, O-Methyl Isoeugenol, 4-(1-Propenyl)pyrocatechol Dimethyl Ether |
PubChem CID | 1549045 |
ChEBI | CHEBI:50550 |
IUPAC Name | 1,2-dimethoxy-4-[(Z)-prop-1-enyl]benzene |
SMILES | CC=CC1=CC(=C(C=C1)OC)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
P110325ML
|
TCI America
P110325ML |
25 mL |
Each for $43.13
|
|
|||||
P1103500ML
|
TCI America
P1103500ML |
500 mL |
Each for $263.78
|
|
|||||
Chemical Identifiers
93-16-3 | |
178.231 | |
NNWHUJCUHAELCL-PLNGDYQASA-N | |
1549045 | |
1,2-dimethoxy-4-[(Z)-prop-1-enyl]benzene |
C11H14O2 | |
MFCD00009282 | |
1,2-Dimethoxy-4-(1-propenyl)benzene, O-Methyl Isoeugenol, 4-(1-Propenyl)pyrocatechol Dimethyl Ether | |
CHEBI:50550 | |
CC=CC1=CC(=C(C=C1)OC)OC |
Specifications
93-16-3 | |
264°C | |
MFCD00009282 | |
1,2-Dimethoxy-4-(1-propenyl)benzene, O-Methyl Isoeugenol, 4-(1-Propenyl)pyrocatechol Dimethyl Ether | |
CC=CC1=CC(=C(C=C1)OC)OC | |
178.231 | |
CHEBI:50550 | |
≥98.0% (GC) | |
4-(1-Propenyl)-1,2-dimethoxybenzene |
Yellow | |
C11H14O2 | |
25 mL | |
NNWHUJCUHAELCL-PLNGDYQASA-N | |
1,2-dimethoxy-4-[(Z)-prop-1-enyl]benzene | |
1549045 | |
178.23 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-0638
RTECSNumber : CZ7000000
TSCA : Yes