missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,13-Diaza-18-crown 6-Ether 98.0+%, TCI America™
$294.52 - $1112.84
Chemical Identifiers
| CAS | 23978-55-4 |
|---|---|
| Molecular Formula | C12H28N2O4 |
| Molecular Weight (g/mol) | 264.37 |
| MDL Number | MFCD00005112 |
| InChI Key | NLMDJJTUQPXZFG-UHFFFAOYSA-P |
| Synonym | kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane |
| PubChem CID | 72805 |
| IUPAC Name | 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane-7,16-diium |
| SMILES | C1COCCOCC[NH2+]CCOCCOCC[NH2+]1 |
Chemical Identifiers
| 23978-55-4 | |
| 264.37 | |
| NLMDJJTUQPXZFG-UHFFFAOYSA-P | |
| 72805 | |
| C1COCCOCC[NH2+]CCOCCOCC[NH2+]1 |
| C12H28N2O4 | |
| MFCD00005112 | |
| kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane | |
| 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane-7,16-diium |
Specifications
| 23978-55-4 | |
| White-Yellow | |
| MFCD00005112 | |
| kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane | |
| C1COCCOCC[NH2+]CCOCCOCC[NH2+]1 | |
| 264.37 | |
| 262.35 | |
| Crystalline Powder |
| 115°C | |
| C12H28N2O4 | |
| 1 g | |
| NLMDJJTUQPXZFG-UHFFFAOYSA-P | |
| 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane-7,16-diium | |
| 72805 | |
| ≥98.0% (T) | |
| 4,13-Diaza-18-crown 6-Ether |
Safety and Handling
TSCA : No