missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(1H-Pyrazol-1-yl)benzoic Acid 98.0+%, TCI America™
Supplier: TCI America P24995G
Specifications
4-(1H-Pyrazol-1-yl)benzoic Acid | |
269°C | |
MFCD02682056 | |
4-1h-pyrazol-1-yl benzoic acid, 4-1-pyrazolyl benzoic acid, benzoic acid, 4-1h-pyrazol-1-yl, 4-pyrazol-1-yl benzoic acid, 4-pyrazol-1-yl-benzoic acid, 4-pyrazolylbenzoic acid, pubchem19583, acmc-1bo47, benzoic acid, 4-pyrazol-1-yl, 4-1h-pyrazole-1-yl benzoic acid | |
C1=CN(N=C1)C2=CC=C(C=C2)C(=O)O | |
188.186 | |
188.19 | |
Crystalline Powder |
16209-00-0 | |
C10H8N2O2 | |
5 g | |
XOEKYPIBVOGCDG-UHFFFAOYSA-N | |
4-pyrazol-1-ylbenzoic acid | |
736527 | |
≥98.0% (GC) |
Chemical Identifiers
16209-00-0 | |
188.186 | |
XOEKYPIBVOGCDG-UHFFFAOYSA-N | |
736527 | |
C1=CN(N=C1)C2=CC=C(C=C2)C(=O)O |
C10H8N2O2 | |
MFCD02682056 | |
4-1h-pyrazol-1-yl benzoic acid, 4-1-pyrazolyl benzoic acid, benzoic acid, 4-1h-pyrazol-1-yl, 4-pyrazol-1-yl benzoic acid, 4-pyrazol-1-yl-benzoic acid, 4-pyrazolylbenzoic acid, pubchem19583, acmc-1bo47, benzoic acid, 4-pyrazol-1-yl, 4-1h-pyrazole-1-yl benzoic acid | |
4-pyrazol-1-ylbenzoic acid |
Safety and Handling
TSCA : No