missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(1H-Pyrrol-1-yl)aniline, 97%, Thermo Scientific™

Chemical Identifiers

CAS 52768-17-9
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD01934575
InChI Key NHLHWHRXMZZWGA-UHFFFAOYSA-N
Synonym 4-1h-pyrrol-1-yl aniline, 4-1-pyrrolyl aniline, 4-pyrrol-1-yl aniline, benzenamine, 4-1h-pyrrol-1-yl, 4-pyrrol-1-yl-phenylamine, 4-pyrrolylphenylamine, 1-p-amino-phenylazole, 4-pyrrol-1-yl-aniline, 1-4-aminophenyl pyrrole, 1-4-amino-phenyl pyrrole
PubChem CID 2795457
IUPAC Name 4-pyrrol-1-ylaniline
SMILES C1=CN(C=C1)C2=CC=C(C=C2)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC25514CB
SDS View Documents
Thermo Scientific Chemicals
CC25514CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC25514DA
SDS View Documents
Thermo Scientific Chemicals
CC25514DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC25514EB
SDS View Documents
Thermo Scientific Chemicals
CC25514EB
25g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

52768-17-9
158.204
NHLHWHRXMZZWGA-UHFFFAOYSA-N
2795457
C1=CN(C=C1)C2=CC=C(C=C2)N
C10H10N2
MFCD01934575
4-1h-pyrrol-1-yl aniline, 4-1-pyrrolyl aniline, 4-pyrrol-1-yl aniline, benzenamine, 4-1h-pyrrol-1-yl, 4-pyrrol-1-yl-phenylamine, 4-pyrrolylphenylamine, 1-p-amino-phenylazole, 4-pyrrol-1-yl-aniline, 1-4-aminophenyl pyrrole, 1-4-amino-phenyl pyrrole
4-pyrrol-1-ylaniline

Specifications

52768-17-9
100.0
Amber Glass Bottle
MFCD01934575
4-1h-pyrrol-1-yl aniline, 4-1-pyrrolyl aniline, 4-pyrrol-1-yl aniline, benzenamine, 4-1h-pyrrol-1-yl, 4-pyrrol-1-yl-phenylamine, 4-pyrrolylphenylamine, 1-p-amino-phenylazole, 4-pyrrol-1-yl-aniline, 1-4-aminophenyl pyrrole, 1-4-amino-phenyl pyrrole
C1=CN(C=C1)C2=CC=C(C=C2)N
158.204
158.2
4-(1H-Pyrrol-1-yl)aniline
96.5
97%
C10H10N2
250mg
NHLHWHRXMZZWGA-UHFFFAOYSA-N
4-pyrrol-1-ylaniline
2795457
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'