missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Benzoxazolyl)-4'-(5-methyl-2-benzoxazolyl)stilbene 95.0+%, TCI America™

$132.35 - $132.35

Chemical Identifiers

CAS 5242-49-9
Molecular Formula C29H20N2O2
Molecular Weight (g/mol) 428.491
InChI Key SOTPOQQKAUOHRO-BQYQJAHWSA-N
PubChem CID 5761588
IUPAC Name 2-[4-[(E)-2-[4-(1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-5-methyl-1,3-benzoxazole
SMILES CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=NC6=CC=CC=C6O5
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B156525G
View Documents
TCI America
B156525G
25 g
Each for $132.35
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5242-49-9
428.491
5761588
CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=NC6=CC=CC=C6O5
C29H20N2O2
SOTPOQQKAUOHRO-BQYQJAHWSA-N
2-[4-[(E)-2-[4-(1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-5-methyl-1,3-benzoxazole

Specifications

5242-49-9
C29H20N2O2
SOTPOQQKAUOHRO-BQYQJAHWSA-N
2-[4-[(E)-2-[4-(1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-5-methyl-1,3-benzoxazole
5761588
≥95.0% (HPLC)
4-(2-Benzoxazolyl)-4′-(5-methyl-2-benzoxazolyl)stilbene
Yellow
25 g
CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=NC6=CC=CC=C6O5
428.491
428.49
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6068

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'