missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(2-furyl)benzaldehyde, Thermo Scientific™

Chemical Identifiers

CAS 60456-77-1
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
InChI Key WBUXKMOCVYRVES-UHFFFAOYSA-N
Synonym 4-2-furyl benzaldehyde, 4-furan-2-yl benzaldehyde, 4-furan-2-yl-benzaldehyde, benzaldehyde,4-2-furanyl, 2-4-formylphenyl furan, 4-2-furanyl benzaldehyde
PubChem CID 2772295
IUPAC Name 4-(furan-2-yl)benzaldehyde
SMILES C1=COC(=C1)C2=CC=C(C=C2)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC34404CB
SDS View Documents
Thermo Scientific Chemicals
CC34404CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC34404DA
SDS View Documents
Thermo Scientific Chemicals
CC34404DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC34404DE
SDS View Documents
Thermo Scientific Chemicals
CC34404DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

60456-77-1
172.183
4-2-furyl benzaldehyde, 4-furan-2-yl benzaldehyde, 4-furan-2-yl-benzaldehyde, benzaldehyde,4-2-furanyl, 2-4-formylphenyl furan, 4-2-furanyl benzaldehyde
4-(furan-2-yl)benzaldehyde
C11H8O2
WBUXKMOCVYRVES-UHFFFAOYSA-N
2772295
C1=COC(=C1)C2=CC=C(C=C2)C=O

Specifications

60456-77-1
250mg
WBUXKMOCVYRVES-UHFFFAOYSA-N
4-(furan-2-yl)benzaldehyde
2772295
97%
C11H8O2
4-2-furyl benzaldehyde, 4-furan-2-yl benzaldehyde, 4-furan-2-yl-benzaldehyde, benzaldehyde,4-2-furanyl, 2-4-formylphenyl furan, 4-2-furanyl benzaldehyde
C1=COC(=C1)C2=CC=C(C=C2)C=O
172.183
172.18
4-(2-furyl)benzaldehyde
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'