missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(2-Hydroxyethyl)pyridine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1378003

 View more versions of this product

Catalog No. AAL1378003


Explore available promotions

Description

Description

4-(2-Hydroxyethyl)pyridine acts as a reagent for the protection of carboxyl groups as their 2-(4-pyridyl)ethyl esters in organic synthesis. Further, it is used as an active pharmaceutical ingredient intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Hydroxyethyl)pyridine acts as a reagent for the protection of carboxyl groups as their 2-(4-pyridyl)ethyl esters in organic synthesis. Further, it is used as an active pharmaceutical ingredient intermediate.

Solubility
Miscible with water.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents, bases and mineral acids.
Specifications

Specifications

4-(2-Hydroxyethyl)pyridine
8°C to 10°C
121°C to 122°C (2 mmHg)
C7H9NO
MFCD00129038
111361
4-pyridineethanol, 2-pyridin-4-yl ethanol, 4-2-hydroxyethyl pyridine, 4-ethanolpyridine, 2-4-pyridyl ethanol, 2-pyridin-4-yl ethan-1-ol, 2-pyridin-4-yl-ethanol, 2-gamma-pyridyl ethanol, unii-i89rdq5yd9
DWPYQDGDWBKJQL-UHFFFAOYSA-N
2-pyridin-4-ylethanol
72921
98%
5344-27-4
1.093
92°C (197°F)
1.538
1 g
Hygroscopic
Miscible with water.
C1=CN=CC=C1CCO
123.155
123.16

Chemical Identifiers

5344-27-4
123.155
DWPYQDGDWBKJQL-UHFFFAOYSA-N
72921
C1=CN=CC=C1CCO
C7H9NO
MFCD00129038
4-pyridineethanol, 2-pyridin-4-yl ethanol, 4-2-hydroxyethyl pyridine, 4-ethanolpyridine, 2-4-pyridyl ethanol, 2-pyridin-4-yl ethan-1-ol, 2-pyridin-4-yl-ethanol, 2-gamma-pyridyl ethanol, unii-i89rdq5yd9
2-pyridin-4-ylethanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 226-286-9

RTECSNumber : UT2971000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only