missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(2-Iodoacetamido)-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 98.0+%, TCI America™
Supplier: TCI America I0487100MG
Specifications
4-(2-Iodoacetamido)-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical | |
127°C | |
MFCD00010171 | |
4-2-iodoacetamido-tempo, 4-2-iodoacetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, n-1-oxyl-2,2,6,6-tetramethyl-4-piperidinyl iodoacetamide, iasl, tmpia, 2,2,6,6-tetramethylpiperdin-n-1-oxyiodoacetamide, n-oxyl-2,2,6,6-tetramethylpiperid-4-yl iodoacetamide, 4-n-iodoacetamide-2,2,6,6-tetramethylpiperidine-1-oxyl, n-1-oxy-2,2,6,6-tetramethyl-4-piperidinyl iodoacetamide, 4-2-iodoacetamido-2,2,6,6-tetramethylpiperidine-1-oxyl | |
CC1(CC(CC(N1[O])(C)C)NC(=O)CI)C | |
339.197 | |
339.20 | |
Crystalline Powder |
25713-24-0 | |
C11H20IN2O2 | |
100 mg | |
UCTVRHAKQRFPEZ-UHFFFAOYSA-N | |
2-iodo-N-(1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-yl)acetamide | |
2724305 | |
≥98.0% (HPLC) |
Chemical Identifiers
25713-24-0 | |
339.197 | |
UCTVRHAKQRFPEZ-UHFFFAOYSA-N | |
2724305 | |
CC1(CC(CC(N1[O])(C)C)NC(=O)CI)C |
C11H20IN2O2 | |
MFCD00010171 | |
4-2-iodoacetamido-tempo, 4-2-iodoacetamido-2,2,6,6-tetramethylpiperidine 1-oxyl, n-1-oxyl-2,2,6,6-tetramethyl-4-piperidinyl iodoacetamide, iasl, tmpia, 2,2,6,6-tetramethylpiperdin-n-1-oxyiodoacetamide, n-oxyl-2,2,6,6-tetramethylpiperid-4-yl iodoacetamide, 4-n-iodoacetamide-2,2,6,6-tetramethylpiperidine-1-oxyl, n-1-oxy-2,2,6,6-tetramethyl-4-piperidinyl iodoacetamide, 4-2-iodoacetamido-2,2,6,6-tetramethylpiperidine-1-oxyl | |
2-iodo-N-(1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-yl)acetamide |
Safety and Handling
TSCA : No