missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(2-Methyl-1H-imidazol-1-yl)aniline, 97%, Thermo Scientific™

Chemical Identifiers

CAS 74852-81-6
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
MDL Number MFCD06797788
InChI Key IEZCMVRWKNEHJB-UHFFFAOYSA-N
Synonym 4-2-methylimidazol-1-yl phenylamine, 4-2-methyl-1h-imidazol-1-yl aniline, 4-2-methyl-imidazol-1-yl-phenylamine, 4-2-methyl-1h-imidazol-1-yl benzenamine, 4-2-methylimidazol-1-yl aniline, 4-2-methyl-1h-imidazol-1-yl phenylamine, benzenamine, 4-2-methyl-1h-imidazol-1-yl, 4-2-methylimidazolyl phenylamine, pubchem19934, 1-4-aminophenyl-2-methylimidazole
PubChem CID 3157478
IUPAC Name 4-(2-methylimidazol-1-yl)aniline
SMILES CC1=NC=CN1C2=CC=C(C=C2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC61414DA
SDS View Documents
Thermo Scientific Chemicals
CC61414DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC61414DE
SDS View Documents
Thermo Scientific Chemicals
CC61414DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

74852-81-6
173.219
IEZCMVRWKNEHJB-UHFFFAOYSA-N
3157478
CC1=NC=CN1C2=CC=C(C=C2)N
C10H11N3
MFCD06797788
4-2-methylimidazol-1-yl phenylamine, 4-2-methyl-1h-imidazol-1-yl aniline, 4-2-methyl-imidazol-1-yl-phenylamine, 4-2-methyl-1h-imidazol-1-yl benzenamine, 4-2-methylimidazol-1-yl aniline, 4-2-methyl-1h-imidazol-1-yl phenylamine, benzenamine, 4-2-methyl-1h-imidazol-1-yl, 4-2-methylimidazolyl phenylamine, pubchem19934, 1-4-aminophenyl-2-methylimidazole
4-(2-methylimidazol-1-yl)aniline

Specifications

74852-81-6
100.0
Amber Glass Bottle
MFCD06797788
4-2-methylimidazol-1-yl phenylamine, 4-2-methyl-1h-imidazol-1-yl aniline, 4-2-methyl-imidazol-1-yl-phenylamine, 4-2-methyl-1h-imidazol-1-yl benzenamine, 4-2-methylimidazol-1-yl aniline, 4-2-methyl-1h-imidazol-1-yl phenylamine, benzenamine, 4-2-methyl-1h-imidazol-1-yl, 4-2-methylimidazolyl phenylamine, pubchem19934, 1-4-aminophenyl-2-methylimidazole
CC1=NC=CN1C2=CC=C(C=C2)N
173.219
173.22
4-(2-Methyl-1H-imidazol-1-yl)aniline
96.5
97%
C10H11N3
1g
IEZCMVRWKNEHJB-UHFFFAOYSA-N
4-(2-methylimidazol-1-yl)aniline
3157478
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'