Learn More
4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3444103
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride | |
104°C to 105°C | |
MFCD00219329 | |
UN3261 | |
4-2-oxopyrrolidin-1-yl benzenesulfonyl chloride, 4-2-oxo-1-pyrrolidinyl benzenesulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonylchloride, 4-2-oxopyrrolidin-1-yl benzene-1-sulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonyl chloride, benzenesulfonylchloride, 4-2-oxo-1-pyrrolidinyl, 1-4-chlorosulfonyl phenyl pyrrolidin-2-one, pubchem5680, acmc-1bri4, benzenesulfonylchloride,4-2-oxo-1-pyrrolidinyl | |
C1CC(=O)N(C1)C2=CC=C(C=C2)S(=O)(=O)Cl | |
259.704 | |
259.71 |
112539-09-0 | |
C10H10ClNO3S | |
1 g | |
Air and moisture sensitive | |
OUAVTZYGHGCCCB-UHFFFAOYSA-N | |
4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride | |
2794593 | |
97% |
Chemical Identifiers
112539-09-0 | |
259.704 | |
OUAVTZYGHGCCCB-UHFFFAOYSA-N | |
4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride |
C10H10ClNO3S | |
MFCD00219329 | |
2794593 | |
C1CC(=O)N(C1)C2=CC=C(C=C2)S(=O)(=O)Cl |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only