missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(2-Tetrahydropyranyloxy)phenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H54115AE

Catalog No. AAH54115AE


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(2-Tetrahydropyranyloxy)phenylmagnesium bromide
C11H13BrMgO2
100 mL
3547417
4-2-tetrahydro-2h-pyranoxy phenylmagnesium bromide, grignard reagent, 2-4-bromomagnesio phenoxy oxane, p-2-tetrahydropyranyloxy phenyl magnesium bromide, p-2-tetrahydropyranyloxy phenyl magnesium bromide, 4-tetrahydropyran-2-yloxy phenyl magnesium bromide, 4-2-tetrahydro-3-h-pyranoxy phenylmagnesium bromide, 4-2-tetrahydro-3-h-pyranoxy phenylmagnesium bromide, 4-2-tetrahydro-2h-pyranoxy phenylmagnesium bromide solution, 4-2-tetrahydro-2h-pyranoxy phenylmagnesium bromide solution, 4-2-tetrahydropyranyloxy phenylmagnesium bromide, 0.5m in 2-methf
Br[Mg]C1=CC=C(OC2CCCCO2)C=C1
281.43
281.43
36637-44-2
MFCD03427254
UN2924
Air and moisture sensitive
OHPUQUBYKZJOLY-UHFFFAOYNA-M
magnesium;2-(phenoxy)oxane;bromide
14519569
0.5M in 2-MeTHF

Chemical Identifiers

36637-44-2
281.43
OHPUQUBYKZJOLY-UHFFFAOYNA-M
14519569
Br[Mg]C1=CC=C(OC2CCCCO2)C=C1
C11H13BrMgO2
MFCD03427254
4-2-tetrahydro-2h-pyranoxy phenylmagnesium bromide, grignard reagent, 2-4-bromomagnesio phenoxy oxane, p-2-tetrahydropyranyloxy phenyl magnesium bromide, p-2-tetrahydropyranyloxy phenyl magnesium bromide, 4-tetrahydropyran-2-yloxy phenyl magnesium bromide, 4-2-tetrahydro-3-h-pyranoxy phenylmagnesium bromide, 4-2-tetrahydro-3-h-pyranoxy phenylmagnesium bromide, 4-2-tetrahydro-2h-pyranoxy phenylmagnesium bromide solution, 4-2-tetrahydro-2h-pyranoxy phenylmagnesium bromide solution, 4-2-tetrahydropyranyloxy phenylmagnesium bromide, 0.5m in 2-methf
magnesium;2-(phenoxy)oxane;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only