Learn More
4-(2-Thienyl)benzenesulfonyl chloride, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H34399MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-(2-Thienyl)benzenesulfonyl chloride | |
107°C to 108°C | |
MFCD11655569 | |
UN3261 | |
IRDZCMUQNGEQEP-UHFFFAOYSA-N | |
4-thiophen-2-ylbenzenesulfonyl chloride | |
18387759 | |
96% |
209592-91-6 | |
C10H7ClO2S2 | |
250 mg | |
4-2-thienyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-2-thienyl, 4-thiophen-2-yl benzene-1-sulfonyl chloride, 4-thiophen-2-yl benzenesulfonyl chloride | |
C1=CSC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl | |
258.734 | |
258.75 |
Chemical Identifiers
209592-91-6 | |
258.734 | |
IRDZCMUQNGEQEP-UHFFFAOYSA-N | |
18387759 | |
C1=CSC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl |
C10H7ClO2S2 | |
MFCD11655569 | |
4-2-thienyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-2-thienyl, 4-thiophen-2-yl benzene-1-sulfonyl chloride, 4-thiophen-2-yl benzenesulfonyl chloride | |
4-thiophen-2-ylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only