missing translation for 'onlineSavingsMsg'
Learn More

4-(2H-Tetrazol-5-yl)benzeneboronic acid pinacol ester, 95%

CAS: 775351-40-1 | C13H17BN4O2 | 272.115 g/mol

$114.74 - $324.82

Chemical Identifiers

CAS 775351-40-1
Molecular Formula C13H17BN4O2
Molecular Weight (g/mol) 272.115
MDL Number MFCD12761127
InChI Key ONAVJWUKLJDJAI-UHFFFAOYSA-N
Synonym 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-2h-tetrazole, 4-1h-tetrazol-5-yl benzene-1-boronic acid pinacol ester, 4-2h-tetrazol-5-yl benzeneboronic acid pinacol ester, 5-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole, 4-2h-tetrazol-5-yl phenylboronic acid pinacol ester, 5-4-phenylboronic acid pinacol ester-2h-tetrazole, 4-1h-tetrazol-5-yl phenyl boronic acid pinacol ester, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-tetrazole, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole
PubChem CID 45785702
IUPAC Name 5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2H-tetrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=NNN=N3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59222MD
View Documents
Thermo Scientific Chemicals
H59222MD
250 mg
Each for $114.74
Only null left
 
AAH5922203
View Documents
Thermo Scientific Chemicals
H5922203
1 g
Each for $324.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

775351-40-1
272.115
ONAVJWUKLJDJAI-UHFFFAOYSA-N
45785702
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=NNN=N3
C13H17BN4O2
MFCD12761127
5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-2h-tetrazole, 4-1h-tetrazol-5-yl benzene-1-boronic acid pinacol ester, 4-2h-tetrazol-5-yl benzeneboronic acid pinacol ester, 5-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole, 4-2h-tetrazol-5-yl phenylboronic acid pinacol ester, 5-4-phenylboronic acid pinacol ester-2h-tetrazole, 4-1h-tetrazol-5-yl phenyl boronic acid pinacol ester, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-tetrazole, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole
5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2H-tetrazole

Specifications

775351-40-1
MFCD12761127
5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-2h-tetrazole, 4-1h-tetrazol-5-yl benzene-1-boronic acid pinacol ester, 4-2h-tetrazol-5-yl benzeneboronic acid pinacol ester, 5-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole, 4-2h-tetrazol-5-yl phenylboronic acid pinacol ester, 5-4-phenylboronic acid pinacol ester-2h-tetrazole, 4-1h-tetrazol-5-yl phenyl boronic acid pinacol ester, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-tetrazole, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-1,2,3,4-tetrazole
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C3=NNN=N3
272.115
272.11
4-(2H-Tetrazol-5-yl)benzeneboronic acid pinacol ester
C13H17BN4O2
250 mg
ONAVJWUKLJDJAI-UHFFFAOYSA-N
5-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2H-tetrazole
45785702
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only