missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one, ≥90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CD02877DA

 View more versions of this product

Catalog No. CD02877DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-(3-Aminopropyl)-2, 4-dihydro-3H-pyrazol-3-one
89.5
90%
C6H11N3O
1g
LMLXMJPJCSUFAB-UHFFFAOYSA-N
4-(3-aminopropyl)-1,4-dihydropyrazol-5-one
2776763
90%
7032-17-9
100.0
Amber Glass Bottle
MFCD00053049
4-3-aminopropyl-2,4-dihydro-3h-pyrazol-3-one, 4-3-aminopropyl-1h-pyrazol-5 4h-one, 4-3-aminopropyl-2,4-dihydropyrazol-3-one, 4-3-aminopropyl-4,5-dihydro-1h-pyrazol-5-one, 3h-pyrazol-3-one,4-3-aminopropyl-2,4-dihydro, aminopropyldihydropyrazolone, 4-3-aminopropyl-2-pyrazolin-5-one, 4-3-aminopropyl-1,4-dihydropyrazol-5-one
C1=NNC(=O)C1CCCN
141.174
141.17

Chemical Identifiers

7032-17-9
141.174
LMLXMJPJCSUFAB-UHFFFAOYSA-N
2776763
C1=NNC(=O)C1CCCN
C6H11N3O
MFCD00053049
4-3-aminopropyl-2,4-dihydro-3h-pyrazol-3-one, 4-3-aminopropyl-1h-pyrazol-5 4h-one, 4-3-aminopropyl-2,4-dihydropyrazol-3-one, 4-3-aminopropyl-4,5-dihydro-1h-pyrazol-5-one, 3h-pyrazol-3-one,4-3-aminopropyl-2,4-dihydro, aminopropyldihydropyrazolone, 4-3-aminopropyl-2-pyrazolin-5-one, 4-3-aminopropyl-1,4-dihydropyrazol-5-one
4-(3-aminopropyl)-1,4-dihydropyrazol-5-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions